• USA Home
  • Search Results

  • potassiumiodide16600768111011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium iodide

19 Products
Image of Potassium iodide
CAS Number:
Linear Formula:
 KI
Molecular Weight:
 166.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Ice Shipping
  Color
  Restriction
  Application(s)
  Potency
  Bulk Density
  Quality clarifier
  Purity Grade
  Loss
  Solubility
  Agency/Method
  Product Line
  Special Grade
  Physical Form
  pH Value
Show More

Product #
Description
221945 Potassium iodide, ACS reagent, ≥99.0%,
30315 Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%,
207969 Potassium iodide, ReagentPlus®, 99%,
746428 Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
60399 Potassium iodide, BioUltra, ≥99.5% (AT),
1.05043 Potassium iodide, for analysis EMSURE® ISO,Reag. Ph Eur,
204102 Potassium iodide, ≥99.99% trace metals basis,
793582 Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%,
03124 Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance),
P8166 Potassium iodide, suitable for plant cell culture,
P2963 Potassium iodide, BioXtra, ≥99.0%,
60422 Potassium iodide, tested according to Ph. Eur.,
1.05040 Potassium iodide, EMPROVE® API Ph Eur,BP,JP,USP,
1.05044 Potassium iodide, 99.995 Suprapur®,
429422 Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis,
PX1505 Potassium iodide, crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
1548280 Potassium iodide, United States Pharmacopeia (USP) Reference Standard,
PX1507 Potassium iodide, granular, Meets ACS Specifications GR ACS,
PHR1360 Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material,