• USA Home
  • Search Results

  • potassiumnitrate10110775779111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium nitrate

15 Products
Image of Potassium nitrate
CAS Number:
Empirical Formula (Hill Notation):
 KNO3
Molecular Weight:
 101.10
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Package quantity
  Manufacturer designation
  Loss
  Agency/Method
  Application(s)
  Product Line
  Potency
  Bulk Density
  Melting Point
  Solubility
  pH Value
  Physical Form
  Special Grade
Show More

Product #
Description
P8394 Potassium nitrate, ReagentPlus®, ≥99.0%,
221295 Potassium nitrate, ACS reagent, ≥99.0%,
P8291 Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture,
31263 Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%,
1.05063 Potassium nitrate, for analysis EMSURE® ISO,Reag. Ph Eur,
542040 Potassium nitrate, 99.999% trace metals basis,
204110 Potassium nitrate, 99.99% trace metals basis,
NIST193 Potassium nitrate, NIST® SRM® 193,
P6083 Potassium nitrate, BioXtra, ≥99.0%,
1.05065 Potassium nitrate, 99.995 Suprapur®,
1.05061 Potassium nitrate, EMPROVE® ESSENTIAL Ph Eur,BP,USP,FCC,E 252,
12648 Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%,
1548349 Potassium nitrate, United States Pharmacopeia (USP) Reference Standard,
PHR2203 Potassium Nitrate, Pharmaceutical Secondary Standard; Certified Reference Material,
PX1520 Potassium Nitrate, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,