• USA Home
  • Search Results

  • potassiumnitrite8510775809011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium nitrite

3 Products
Image of Potassium nitrite
CAS Number:
Empirical Formula (Hill Notation):
 KNO2
Molecular Weight:
 85.10
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
Show More