• USA Home
  • Search Results

  • potassiumoxalatemonohydrate18423648748511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium oxalate monohydrate

2 Products
Synonym:   Ethanedioic acid, Oxalic acid potassium salt
Image of Potassium oxalate monohydrate
CAS Number:
Linear Formula:
 (COOK)2 · H2O
Molecular Weight:
 184.23
 Beilstein/REAXYS Number:
 3752576
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Solubility
  Product Line
  Physical Form
  Special Grade
  pH Value
Show More