• USA Home
  • Search Results

  • potassiumperchlorate13855777874711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium perchlorate

3 Products
Image of Potassium perchlorate
CAS Number:
Empirical Formula (Hill Notation):
 ClKO4
Molecular Weight:
 138.55
 EC Index Number
 231-912-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More