• USA Home
  • Search Results

  • potassiumpermanganate15803772264711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium permanganate

10 Products
Image of Potassium permanganate
CAS Number:
Empirical Formula (Hill Notation):
 KMnO4
Molecular Weight:
 158.03
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Suitability
  Product Line
  Loss
  Vapor pressure
  Potency
  Melting Point
  Density
  Bulk Density
  Restriction
  Agency/Method
  Purity Grade
  Solubility
  Special Grade
  pH Value
  Assay
Show More