• USA Home
  • Search Results

  • potassiumphosphatedibasic17418775811411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium phosphate dibasic

9 Products
Synonym:   Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate
Image of Potassium phosphate dibasic
CAS Number:
Linear Formula:
 K2HPO4
Molecular Weight:
 174.18
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38070212
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Quality clarifier
  Purity Grade
  Physical Form
  Solubility
  Loss
  UV-Absorption Values
  pH Value
  Assay
Show More