• USA Home
  • Search Results

  • potassiumphosphatedibasicsolution17418775811411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium phosphate dibasic solution

1 Products
Synonym:   Dipotassium phosphate
Image of Potassium phosphate dibasic solution
CAS Number:
Empirical Formula (Hill Notation):
 HK2O4P
Molecular Weight:
 174.18
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More