• USA Home
  • Search Results

  • potassiumphosphatemonobasic13609777877011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium phosphate monobasic

16 Products
Synonym:   Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate
Image of Potassium phosphate monobasic
CAS Number:
Linear Formula:
 KH2PO4
Molecular Weight:
 136.09
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  UV-Absorption Condition
  Ice Shipping
  Restriction
  Quality clarifier
  Purity Grade
  Application(s)
  Physical Form
  UV-Absorption Values
  Loss
  pH Value
  Solubility
  Assay
  pK-Values
Show More

Product #
Description
P5655 Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%,
229806 Potassium phosphate monobasic, 99.99% trace metals basis,
P9791 Potassium phosphate monobasic, for molecular biology, ≥98.0%,
P0662 Potassium phosphate monobasic, ACS reagent, ≥99.0%,
P5379 Potassium phosphate monobasic, ReagentPlus®,
04243 Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance),
60220 Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%,
60218 Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T),
60229 Potassium phosphate monobasic, tested according to Ph Eur, anhydrous,
795488 Potassium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
57618 Potassium phosphate monobasic, for HPLC, LiChropur, 99.5-101.0% (T),
RDD020 Potassium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.0% (titration),
92214 Potassium phosphate monobasic, TraceCERT®, certified reference material, 31P-qNMR Standard,
NIST2186I Potassium phosphate monobasic, NIST® SRM® 2186I,
PX1562 Potassium phosphate monobasic, HPLC, Meets ACS Specifications,
PX1565 Potassium phosphate monobasic, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,