• USA Home
  • Search Results

  • potassiumphthalatemonobasic2042287724711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium phthalate monobasic

7 Products
Synonym:   KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate
Image of Potassium phthalate monobasic
CAS Number:
Linear Formula:
 HOOCC6H4COOK
Molecular Weight:
 204.22
 Beilstein/REAXYS Number:
 3637128
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Anal. Standard Form
  Solubility
  Physical Form
  Application(s)
  Quality clarifier
  Loss
  UV-Absorption Values
  Purity Grade
  Special Grade
  pH Value
Show More