• USA Home
  • Search Results

  • potassiumsorbate150222463461511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium sorbate

8 Products
Synonym:   Potassium sorbate, Sorbic acid potassium salt
Image of Potassium sorbate
CAS Number:
Empirical Formula (Hill Notation):
 C6H7KO2
Molecular Weight:
 150.22
 E Number:
 E202
 EC Index Number
 246-376-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  pH Value
  Vapor pressure
  Solubility
  Purity Grade
  Potency
  Melting Point
  Density
  Bulk Density
  Autoignition temp
  Assay
  Anal. Standard Form
  Physical Form
  Titertest
Show More