• USA Home
  • Search Results

  • potassiumtertpentoxidesolution126244123393611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium tert-pentoxide solution

2 Products
Synonym:   Potassium (2-methyl-2-butoxide), Potassium tert-pentylate
Image of Potassium <I>tert</I>-pentoxide solution
CAS Number:
Linear Formula:
 CH3CH2C(CH3)2OK
Molecular Weight:
 126.24
 Beilstein/REAXYS Number:
 4149025
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More