• USA Home
  • Search Results

  • potassiumtetraboratetetrahydrate305501204578211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium tetraborate tetrahydrate

2 Products
Synonym:   Potassium biborate
Image of Potassium tetraborate tetrahydrate
CAS Number:
Linear Formula:
 K2B4O7 · 4H2O
Molecular Weight:
 305.50
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More