• USA Home
  • Search Results

  • potassiumtetrachloropalladateii326431002598611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium tetrachloropalladate(II)

2 Products
Synonym:   Potassium palladium(II) chloride
Image of Potassium tetrachloropalladate(II)
CAS Number:
Linear Formula:
 K2PdCl4
Molecular Weight:
 326.43
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More