• USA Home
  • Search Results

  • potassiumtetrakis4chlorophenylborate496111468077411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium tetrakis(4-chlorophenyl)borate

1 Products
Synonym:   Tetrakis(4-chlorophenyl)boron potassium
Image of Potassium tetrakis(4-<WBR>chlorophenyl)<WBR>borate
CAS Number:
Linear Formula:
 (ClC6H4)4BK
Molecular Weight:
 496.11
 Beilstein/REAXYS Number:
 4117827
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More