• USA Home
  • Search Results

  • potassiumthiocyanate971833320011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium thiocyanate

5 Products
Synonym:   Potassium rhodanide
Image of Potassium thiocyanate
CAS Number:
Linear Formula:
 KSCN
Molecular Weight:
 97.18
 Beilstein/REAXYS Number:
 3594799
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Vapor pressure
  Solubility
  Potency
  Melting Point
  Density
  Bulk Density
  Agency/Method
  Product Line
  Special Grade
  Physical Form
  pH Value
Show More