• USA Home
  • Search Results

  • potassiumthiocyanatesolution971833320011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium thiocyanate solution

1 Products
Synonym:   Potassium rhodanide
Image of Potassium thiocyanate solution
CAS Number:
Linear Formula:
 KSCN
Molecular Weight:
 97.18
 Beilstein/REAXYS Number:
 3594799
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  pH Value
Show More