• USA Home
  • Search Results

  • praseodymium14091744010011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Praseodymium

28 Products
Image of Praseodymium
CAS Number:
Empirical Formula (Hill Notation):
 Pr
Molecular Weight:
 140.91
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Weight
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Physical Form
Show More

Product #
Description
263176 Praseodymium, powder, ~40 mesh, 99.5% trace rare earth metals basis,
261173 Praseodymium, ingot, under oil, 99.9% trace rare earth metals basis,
GF67194018 Praseodymium, lump, 25 mm max. lump size, weight 100 g, purity 99%,
GF77174062 Praseodymium, foil, thickness 0.50 mm, size 50 × 50 mm, purity 99%,
GF81348532 Praseodymium, rod, 10.0 mm diameter, length 25 mm, purity 99%,
GF89418606 Praseodymium, foil, thickness 1.0 mm, size 50 × 50 mm, purity 99%,
GF99251590 Praseodymium, lump, 25 mm max. lump size, weight 200 g, purity 99%,
GF03436141 Praseodymium, foil, thickness 0.50 mm, size 25 × 25 mm, purity 99%,
GF06449036 Praseodymium, lump, 25 mm max. lump size, weight 300 g, purity 99%,
GF15580098 Praseodymium, powder, max. particle size 250 micron, weight 10 g, purity 99.9%,
GF17510600 Praseodymium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99%,
GF38937550 Praseodymium, powder, max. particle size 250 micron, weight 50 g, purity 99.9%,
GF47511503 Praseodymium, powder, max. particle size 250 micron, weight 20 g, purity 99.9%,
GF52374225 Praseodymium, rod, 10.0 mm diameter, length 50 mm, purity 99%,
GF91191191 Praseodymium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%,
GF98899687 Praseodymium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%,
GF96026531 Praseodymium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%,
GF75299170 Praseodymium, foil, not light tested, 25x25mm, thickness 0.015mm, as rolled, 99%,
GF69747621 Praseodymium, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99%,
GF82608295 Praseodymium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%,
GF34755857 Praseodymium, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99%,
GF92630246 Praseodymium, foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%,
GF48004804 Praseodymium, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99%,
GF66110329 Praseodymium, rod, 100mm, diameter 6.35mm, cast, 99%,
GF06128883 Praseodymium, rod, 50mm, diameter 6.35mm, cast, 99%,
GF50132191 Praseodymium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%,
GF66524643 Praseodymium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%,
GF07601059 Praseodymium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%,