• USA Home
  • Search Results

  • praseodymiumiiinitratehexahydrate435011587877011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Praseodymium(III) nitrate hexahydrate

1 Products
Synonym:   Praseodymium trinitrate hexahydrate
Image of Praseodymium(III) nitrate hexahydrate
CAS Number:
Linear Formula:
 Pr(NO3)3·6H2O
Molecular Weight:
 435.01
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More