• USA Home
  • Search Results

  • praziquantel312415526874111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Praziquantel

5 Products
Synonym:   2-(Cyclohexylcarbonyl)-1,2,3,6,7-11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Praziquantel
Image of Praziquantel
CAS Number:
Empirical Formula (Hill Notation):
 C19H24N2O2
Molecular Weight:
 312.41
 Beilstein/REAXYS Number:
 761557
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Mode of Action
  Activity Spectrum
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Application(s)
Show More