• USA Home
  • Search Results

  • prednisolone360445024811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Prednisolone

6 Products
Synonym:   1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrohydrocortisone, 1-Dehydrocortisol, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione
Image of Prednisolone
CAS Number:
Empirical Formula (Hill Notation):
 C21H28O5
Molecular Weight:
 360.44
 Beilstein/REAXYS Number:
 1354103
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Manufacturer designation
  Physical Form
  Application(s)
Show More