• USA Home
  • Search Results

  • progesterone314465783011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Progesterone

9 Products
Synonym:   4-Pregnene-3,20-dione
Image of Progesterone
CAS Number:
Empirical Formula (Hill Notation):
 C21H30O2
Molecular Weight:
 314.46
 Beilstein/REAXYS Number:
 1915950
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Potency
  Ice Shipping
  ID clarifier
  Solubility
  Manufacturer designation
  Biological Source
  Anal. Standard Form
  Product Line
  Application(s)
  Assay
  Physical Form
Show More