• USA Home
  • Search Results

  • propafenonehydrochloride377903418322711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propafenone hydrochloride

3 Products
Synonym:   1-[2-(2-Hydroxy-3-(propylamino)propoxy)phenyl]-3-phenyl-1-propanone hydrochloride
Image of Propafenone hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H27NO3 · HCl
Molecular Weight:
 377.90
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More