• USA Home
  • Search Results

  • propargylalcohol560610719711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propargyl alcohol

1 Products
Synonym:   2-Propyn-1-ol
Image of Propargyl alcohol
CAS Number:
Linear Formula:
 HC≡CCH2OH
Molecular Weight:
 56.06
 Beilstein/REAXYS Number:
 506003
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P50803 Propargyl alcohol, 99%,