• USA Home
  • Search Results

  • propargylamine5508245071711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propargylamine

1 Products
Synonym:   2-Propynylamine, 3-Amino-1-propyne
Image of Propargylamine
CAS Number:
Linear Formula:
 HC≡CCH2NH2
Molecular Weight:
 55.08
 Beilstein/REAXYS Number:
 773681
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
P50900 Propargylamine, 98%,