• USA Home
  • Search Results

  • propargylaminehydrochloride91541543052111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propargylamine hydrochloride

1 Products
Synonym:   2-Propynylamine hydrochloride, 3-Amino-1-propyne hydrochloride
Image of Propargylamine hydrochloride
CAS Number:
Linear Formula:
 HC≡CCH2NH2 · HCl
Molecular Weight:
 91.54
 Beilstein/REAXYS Number:
 3669858
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More