• USA Home
  • Search Results

  • propargylbromidesolution1189610696711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propargyl bromide solution

3 Products
Synonym:   3-Bromo-1-propyne
Image of Propargyl bromide solution
CAS Number:
Linear Formula:
 HC≡CCH2Br
Molecular Weight:
 118.96
 Beilstein/REAXYS Number:
 605309
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Additive
  Density
Show More