• USA Home
  • Search Results

  • propionicacid74087909411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propionic acid

9 Products
Synonym:   Acid C3, Propanoic acid, Propanyl acid
Image of Propionic acid
CAS Number:
Linear Formula:
 CH3CH2COOH
Molecular Weight:
 74.08
 Beilstein/REAXYS Number:
 506071
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39021305
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Color
  Document Links
  Allergen
  Application(s)
  Refractive Index
  Special Grade
  Organoleptic property
  Density
  Biological Source
  Agency/Method
  Solubility
  Assay
Show More