• USA Home
  • Search Results

  • propionicanhydride1301412362611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propionic anhydride

4 Products
Synonym:   Propanoic anhydride
Image of Propionic anhydride
CAS Number:
Linear Formula:
 (CH3CH2CO)2O
Molecular Weight:
 130.14
 Beilstein/REAXYS Number:
 507066
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Potency
  Density
  Assay
Show More