• USA Home
  • Search Results

  • propionitrile550810712011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propionitrile

4 Products
Synonym:   Ethyl cyanide
Image of Propionitrile
CAS Number:
Linear Formula:
 C₃H₅N
Molecular Weight:
 55.08 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Titertest
  Refractive Index
  Assay
Show More