• USA Home
  • Search Results

  • propofol17827207854811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propofol

4 Products
Synonym:   2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, 2,6-Diisopropylphenol, Propofol
Image of Propofol
CAS Number:
Linear Formula:
 [(CH3)2CH]2C6H3OH
Molecular Weight:
 178.27
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More