• USA Home
  • Search Results

  • propyl4hydroxybenzoate180209413311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl 4-hydroxybenzoate

5 Products
Synonym:   4-Hydroxybenzoic acid propyl ester, Propyl paraben
Image of Propyl 4-hydroxybenzoate
CAS Number:
Linear Formula:
 HOC6H4CO2CH2CH2CH3
Molecular Weight:
 180.20
 Beilstein/REAXYS Number:
 1103245
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Solubility
  Product Line
  Special Grade
  Assay
Show More