• USA Home
  • Search Results

  • propylacetate1021310960411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl acetate

9 Products
Image of Propyl acetate
CAS Number:
Linear Formula:
 CH3COOCH2CH2CH3
Molecular Weight:
 102.13
 Beilstein/REAXYS Number:
 1740764
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39022103
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Document Links
  Anal. Standard Form
  Allergen
  Organoleptic property
  Solubility
  Color
  Application(s)
  Package quantity
  Physical Form
  Agency/Method
  Assay
Show More