• USA Home
  • Search Results

  • propylamine591110710811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propylamine

5 Products
Synonym:   1-Aminopropane
Image of Propylamine
CAS Number:
Linear Formula:
 CH3CH2CH2NH2
Molecular Weight:
 59.11
 Beilstein/REAXYS Number:
 1098243
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39030103
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Refractive Index
  Titertest
  Potency
  Assay
Show More