• USA Home
  • Search Results

  • propylbenzene1201910365111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propylbenzene

4 Products
Synonym:   1-Phenylpropane, Isocumene
Image of Propylbenzene
CAS Number:
Linear Formula:
 C6H5CH2CH2CH3
Molecular Weight:
 120.19
 Beilstein/REAXYS Number:
 1903006
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Refractive Index
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More