• USA Home
  • Search Results

  • propylbutyrate1301810566811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl butyrate

3 Products
Synonym:   n-Propyl-n-butanoate
Image of Propyl butyrate
CAS Number:
Linear Formula:
 CH3CH2CH2COOCH2CH2CH3
Molecular Weight:
 130.18
 Beilstein/REAXYS Number:
 1745552
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Assay
Show More