• USA Home
  • Search Results

  • propylenecarbonate1020910832711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propylene carbonate

7 Products
Synonym:   1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one
Image of Propylene carbonate
CAS Number:
Empirical Formula (Hill Notation):
 C4H6O3
Molecular Weight:
 102.09
 Beilstein/REAXYS Number:
 107913
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
  Physical Form
  UV-Absorption Values
  pH Value
  Special Grade
Show More