• USA Home
  • Search Results

  • propylformate881111074711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl formate

2 Products
Synonym:   n-Propyl methanoate
Image of Propyl formate
CAS Number:
Linear Formula:
 HCOOCH2CH2CH3
Molecular Weight:
 88.11
 Beilstein/REAXYS Number:
 1739272
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  Assay
Show More