• USA Home
  • Search Results

  • propylgallate2122012179911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl gallate

6 Products
Synonym:   3,4,5-Trihydroxybenzoic acid propyl ester
Image of Propyl gallate
CAS Number:
Linear Formula:
 3,4,5-(HO)3C6H2CO2CH2CH2CH3
Molecular Weight:
 212.20
 Beilstein/REAXYS Number:
 1877976
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 39024506
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Manufacturer designation
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
  Special Grade
Show More