• USA Home
  • Search Results

  • propylisovalerate1442155700611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl isovalerate

1 Products
Synonym:   Propyl 3-methylbutanoate, Propyl 3-methylbutyrate
Image of Propyl isovalerate
CAS Number:
Linear Formula:
 (CH3)2CHCH2COO(CH2)2CH3
Molecular Weight:
 144.21
 Beilstein/REAXYS Number:
 1747146
 Council of Europe no.:
 443
 EC Number:
 FEMA Number:
 2960
 Flavis number:
 9.448
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More