• USA Home
  • Search Results

  • propylparaben180209413311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propylparaben

3 Products
Synonym:   4-Hydroxybenzoic acid propyl ester, Propyl 4-hydroxybenzoate, Propyl paraben
Image of Propylparaben
CAS Number:
Linear Formula:
 HOC6H4CO2CH2CH2CH3
Molecular Weight:
 180.20
 Beilstein/REAXYS Number:
 1103245
 E Number:
 E216
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More