• USA Home
  • Search Results

  • propylpropionate1161610636511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Propyl propionate

2 Products
Image of Propyl propionate
CAS Number:
Linear Formula:
 CH3CH2COOCH2CH2CH3
Molecular Weight:
 116.16
 Beilstein/REAXYS Number:
 1699993
 Council of Europe no.:
 403
 EC Number:
 FEMA Number:
 2958
 Flavis number:
 9.122
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Special Grade
  Assay
  Agency/Method
Show More