• USA Home
  • Search Results

  • prostaglandinb2334451336785611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Prostaglandin B2

1 Products
Synonym:   (5Z,13E,15S)-15-Hydroxy-9-oxoprosta-5,8(12),13-trien-1-oic acid, PGB2
Image of Prostaglandin B<SUB>2</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C20H30O4
Molecular Weight:
 334.45
 Beilstein/REAXYS Number:
 2153006
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More