• USA Home
  • Search Results

  • proteasefromstreptomycesgriseus12345903606011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Protease from Streptomyces griseus

2 Products
Synonym:   Actinase E, Pronase E
CAS Number:
 EC Number:
 MDL number:
 eCl@ss:
 32160410
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Solubility
  Product Line
Show More