• USA Home
  • Search Results

  • proteomassinsulinchainboxidizedmaldimsstandard3495893000372611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ProteoMass Insulin chain B oxidized MALDI-MS Standard

1 Products
Synonym:   Insulin Chain B Oxidized from bovine pancreas
CAS Number:
Empirical Formula (Hill Notation):
 C157H232N40O47S2
Molecular Weight:
 3495.89
 MDL number:
 NACRES:
 NA.24
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More