• USA Home
  • Search Results

  • ptolualdehyde1201510487011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Tolualdehyde

3 Products
Synonym:   4-Methylbenzaldehyde
Image of <I>p</I>-Tolualdehyde
CAS Number:
Linear Formula:
 CH3C6H4CHO
Molecular Weight:
 120.15
 Beilstein Registry Number:
 385772
 Council of Europe no.:
 115
 EC Number:
 FEMA Number:
 3068
 Flavis number:
 5.029
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Assay
Show More