• USA Home
  • Search Results

  • ptoluenesulfonamide171227055311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Toluenesulfonamide

4 Products
Image of <I>p</I>-Toluenesulfonamide
CAS Number:
Linear Formula:
 CH3C6H4SO2NH2
Molecular Weight:
 171.22
 Beilstein/REAXYS Number:
 472689
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Assay
Show More