• USA Home
  • Search Results

  • ptoluenesulfonicacidmonohydrate19022619252511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Toluenesulfonic acid monohydrate

4 Products
Image of <I>p</I>-Toluenesulfonic acid monohydrate
CAS Number:
Linear Formula:
 CH3C6H4SO3H · H2O
Molecular Weight:
 190.22
 Beilstein/REAXYS Number:
 3568023
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
  Restriction
  Product Line
  Special Grade
Show More