• USA Home
  • Search Results

  • ptoluidine1071510649011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Toluidine

6 Products
Synonym:   4-Aminotoluene, 4-Methylaniline
Image of <I>p</I>-Toluidine
CAS Number:
Linear Formula:
 CH3C6H4NH2
Molecular Weight:
 107.15
 Beilstein/REAXYS Number:
 471281
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  pH Value
  Transition temperature
  Application(s)
  Potency
  Physical Form
  Vapor pressure
  Assay
Show More