• USA Home
  • Search Results

  • ptolylacetate1501714039611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Tolyl acetate

2 Products
Synonym:   p-Cresyl acetate
Image of <I>p</I>-Tolyl acetate
CAS Number:
Linear Formula:
 CH3CO2C6H4CH3
Molecular Weight:
 150.17
 Council of Europe no.:
 226
 EC Number:
 FEMA Number:
 3073
 Flavis number:
 9.036
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More